
4-Piperidone monohydrate hydrochloride ≥98.0% (by titrimetric analysis)
: TCI
:40064-34-4
:C₅H₉NO·xHCl·yH₂O
:MFCD00012776
:99.13 g/mol (free base)
:254-779-9
:GHS07
- Appearance:White to Yellow to Orange powder to crystaline
- Purity (Argentometric Titration):min. 98.0 %
- Water:11.0 to 13.0 %