
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine Hydrochloride ≥98.0% (by HPLC, total nitrogen) for oxime preparation
Supplier: TCI
Certificates
About this item
CAS Number:57981-02-9
:
Formula:C₇H₄F₅NO
MDL Number:MFCD00012953
:249.57 g/mol
Melting Pt:215°C
EINECS:261-057-7
:GHS07
Specification Test Results
- Appearance:White powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together