Order Entry
Finland
ContactUsLinkComponent
2-Propylcyclohexanol (cis- and trans- mixture) cis- and trans-mixture ≥90.0% (by GC)
2-Propylcyclohexanol (cis- and trans- mixture) cis- and trans-mixture ≥90.0% (by GC)
Supplier:  TCI
Certificates
2-Propylcyclohexanol (cis- and trans- mixture) cis- and trans-mixture ≥90.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 90.0 % (total of isomers)

Specification Test Results

Related Information

Frequently Bought Together