2-Oxotetrahydrofuran-3-yl methacrylate ≥98.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:195000-66-9
:
Formula:C₈H₁₀O₄
MDL Number:MFCD19689400
:170.16 g/mol
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %