Sodium tricyanomethanide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:36603-80-2
:
Formula:C₄N₃Na
MDL Number:MFCD00174290
:113.05 g/mol
UN:3439
ADR:6.1,III
:GHS05
:GHS06
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Ion exchange titration):min. 98.0 %