Order Entry
Finland
ContactUsLinkComponent
2-n-Octyl-1-dodecylamine ≥97.0% (by GC, titration analysis)
Corrosive
2-n-Octyl-1-dodecylamine ≥97.0% (by GC, titration analysis)
Catalog # TCIAO0435-1G
Supplier:  TCI
CAS Number:  
Corrosive
2-n-Octyl-1-dodecylamine ≥97.0% (by GC, titration analysis)
Catalog # TCIAO0435-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 97.0 %
  • Purity (Neutralization titration):
    min. 97.0 %

Specification Test Results

About this item

 :
 :297.57 g/mol
UN:2735
ADR:8,II
 :GHS05
CAS Number:62281-06-5
Formula:C₂₀H₄₃N
MDL Number:MFCD19440743