7-Methoxy-8-(3-methyl-2-buten-1-yl)-2H-chromen-2-one ≥98.0% (by GC)
: TCI
:484-12-8
:
:C₁₅H₁₆O₃
:MFCD00076049
:244.2857 g/mol
:14,06895
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:82.0 to 86.0 deg-C