Isoamyl n-octanoate (contains 2-Methylbutyl n-Octanoate)
TCIAO0031-25ML
: TCI
:
- Pk:25 mL
- Seq:2315
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 % (total of isomer)
:
:214.35 g/mol
:250°C
:85°C
:218-004-8
:2035-99-6
:C₁₃H₂₆O₂
:MFCD00048917