Order Entry
Finland
ContactUsLinkComponent
Nicotinic acid (Niacin) ≥99.5% (by HPLC, titration analysis) matrix for MALDI-TOF/MS
Irritant
Nicotinic acid (Niacin) ≥99.5% (by HPLC, titration analysis) matrix for MALDI-TOF/MS
Supplier:  TCI
Certificates
undefined
Irritant
Nicotinic acid (Niacin) ≥99.5% (by HPLC, titration analysis) matrix for MALDI-TOF/MS
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 99.5 area%
  • Purity (Neutralization titration):
    min. 99.5 %
  • Melting point:
    236.0 to 240.0 deg-C

Specification Test Results