4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:3459-18-5
:
Formula:C₁₄H₁₈N₂O₈
MDL Number:MFCD00063696
:342.30 g/mol
Melting Pt:205°C
Storage Temperature:Freezer
EINECS:222-398-7
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Specific rotation [a]20/D:-12.0 to -17.0 deg (C=0.25, H2O)