4-Nitro-D-phenylalanine hydrate ≥97.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:56613-61-7
Formula:C₉H₁₀N₂O₄·H₂O
MDL Number:MFCD00150962
:228.202 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 97.0 % (calcd.on anh.substance)
- Specific rotation [a]20/D:-5.0 to -8.0 deg (C=2, 1mol/L HCl)
- Water:max. 10.0 %