
4-(4-Nitrophenylazo)phenol ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1435-60-5
:
Formula:C₁₂H₉N₃O₃
MDL Number:MFCD00161165
:243.22 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %