
1,2-Naphthalic anhydride ≥98.0% (by GC, titration analysis)
: TCI
:5343-99-7
:
:C₁₂H₆O₃
:MFCD00191561
:198.18 g/mol
:GHS07
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:168.0 to 172.0 deg-C