Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Zinc naphthenate
Zinc naphthenate (∼7% Zn)
Zinc naphthenate (∼7% Zn)
Zinc naphthenate (∼7% Zn)
TCIAN0341-25G
:
TCI
:
Zinc naphthenate (∼7% Zn)
TCIAN0341-25G
:
TCI
:
Pk:
25 g
Seq:
3308
Appearance:
Colorless to Brown clear liquid
Zinc:
9.5 to 10.5 %
:
:
319.71 g/mol
:
234-409-2
:
3082
:
9,III
:
GHS07
:
GHS09
:
12001-85-3
:
2(C₁₁H₇O₂)·Zn
:
MFCD00147699