4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP) ≥98.0% (by HPLC, titration analysis)
: TCI
:333338-18-4
:4264-83-9
:C₆H₄NNa₂O₆P·6H₂O
:MFCD00066288
:371.144 g/mol
:Refrigerator
:224-246-5
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Ion exchange titration):min. 98.0 % (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:28.0 to 30.0 %