
2-Methyl-5-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
TCIAM3020-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Light yellow powder to lump
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
:
:219.0878 g/mol
:GHS07
:Refrigerator
:610768-32-6
:C₁₂H₁₈BNO₂
:MFCD08669608