Order Entry
Finland
ContactUsLinkComponent
[2-(2-Methoxyethoxy)ethoxy]acetic acid(3,6,9-trioxadecanoic acid) ≥95.0% (by GC, titration analysis)
Irritant
[2-(2-Methoxyethoxy)ethoxy]acetic acid(3,6,9-trioxadecanoic acid) ≥95.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
[2-(2-Methoxyethoxy)ethoxy]acetic acid(3,6,9-trioxadecanoic acid) ≥95.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %
  • Purity (Neutralization titration):
    min. 95.0 %

Specification Test Results