
2-Mercaptopyrazine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAM2950-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
About this item
:
:112.15 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:38521-06-1
Formula:C₄H₄N₂S
MDL Number:MFCD00235012