Methyl-1-(tert-butoxycarbonyl)-piperidine-2-carboxylate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:167423-93-0
:
Formula:C₁₂H₂₁NO₄
MDL Number:MFCD06795925
:243.31 g/mol
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Acid Value:max. 2.0