Order Entry
Finland
ContactUsLinkComponent
3-Methyl-1,2-cyclopentanedione ≥98.0% (by GC, titration analysis)
3-Methyl-1,2-cyclopentanedione ≥98.0% (by GC, titration analysis)
Catalog # TCIAM2904-25G
Supplier:  TCI
CAS Number:  
undefined
3-Methyl-1,2-cyclopentanedione ≥98.0% (by GC, titration analysis)
Catalog # TCIAM2904-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    104.0 to 108.0 deg-C

Specification Test Results

About this item

 :
 :112.13 g/mol
EINECS:212-154-8
CAS Number:765-70-8
Formula:C₆H₈O₂
MDL Number:MFCD00001417