
trans-2-Methyl-2-pentenoic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:16957-70-3
:
Formula:C₆H₁₀O₂
MDL Number:MFCD00002655
:114.14 g/mol
Boiling Pt:125°C
Flash Pt:108°C
EINECS:241-026-4
UN:3265
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %