EMCH.TFA (E-Maleimidocaproic acid hydrazine trifluoroacetic acid salt) ≥95.0% (by HPLC)
Catalog # TCIAM2735-250MG
Supplier: TCI
CAS Number:
Specifications
- Pk:250 mg
- Seq:2590
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Elemental analysis (Nitrogen):11.50 to 13.00 %
Specification Test Results
About this item
:
:339.27 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:151038-94-7
:175290-73-0
Formula:C₁₀H₁₅N₃O₃·C₂HF₃O₂
MDL Number:MFCD07357647