Order Entry
Finland
ContactUsLinkComponent
4-Methoxy-m-toluic acid ≥98.0% (by GC, titration analysis)
4-Methoxy-m-toluic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
4-Methoxy-m-toluic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystaline
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    195.0 to 199.0 deg-C

Specification Test Results

Frequently Bought Together