Methyl (Phenyl 5-Acetamido-4,7,8-tri-O-acetyl-9-azido-3,5,9-trideoxy-2-thio-D-glycero-beta-D-galacto-2-nonulopyranosid)onate ≥92.0% (by HPLC)
TCIAM2695-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 92.0 area%
- Elemental analysis (Nitrogen):9.40 to 10.40 %
:
:566.58 g/mol
:Refrigerator
:219814-65-0
:C₂₄H₃₀N₄O₁₀S