
1-Methyl-1H-pyrazole-5-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Catalog # TCIAM2681-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:69.0 to 73.0 deg-C
Specification Test Results
About this item
:
:208.07 g/mol
:GHS07
CAS Number:847818-74-0
Formula:C₁₀H₁₇BN₂O₂
MDL Number:MFCD05861380