
2-Methoxy-5-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:445264-61-9
:
Formula:C₁₂H₁₈BNO₃
MDL Number:MFCD05663858
:235.09 g/mol
Boiling Pt:102°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:51.0 to 55.0 deg-C