
3-Cyano-6-methylchromone ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:50743-18-5
:
Formula:C₁₁H₇NO₂
MDL Number:MFCD00191962
:185.19 g/mol
UN:3439
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:150.0 to 154.0 deg-C