
1-Methoxy-4-methylcyclohexane (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:90200-72-9
:
Formula:C₈H₁₆O
MDL Number:MFCD27966267
Boiling Pt:148°C
UN:3271
ADR:3,III
:128.22 g/mol
:GHS02
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 % (total of isomers)
Specification Test Results
Frequently Bought Together