3-Methylflavone-8-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3468-01-7
:
Formula:C₁₇H₁₂O₄
MDL Number:MFCD00040998
:280.28 g/mol
EINECS:222-425-2
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:234.0 to 238.0 deg-C