


Methyl-4-(bromomethyl)-3-methoxybenzoate ≥97.0% (by GC)
: TCI
:70264-94-7
:
:C₁₀H₁₁BrO₃
:MFCD00270115
:259.0965 g/mol
:410-310-1
:3077
:9,III
:GHS05
:GHS07
:GHS09
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:92.0 to 96.0 deg-C