Methyl trans,trans-9,12-octadecadienoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2566-97-4
:
Formula:C₁₉H₃₄O₂
MDL Number:MFCD00069997
:294.48 g/mol
Boiling Pt:152°C
Storage Temperature:Freezer
EINECS:219-901-7
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %