Methyl-4-(aminomethyl)benzoate hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:6232-11-7
:
Formula:C₉H₁₁NO₂
MDL Number:MFCD00182671
:201.65 g/mol
Melting Pt:238°C
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together