Order Entry
Finland
ContactUsLinkComponent
N-Methoxy-N-methylcarbamoyl chloride ≥98.0% (by GC, titration analysis)
Corrosive
N-Methoxy-N-methylcarbamoyl chloride ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Corrosive
N-Methoxy-N-methylcarbamoyl chloride ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %

Specification Test Results