Order Entry
Finland
ContactUsLinkComponent
8-Mercaptomenthone ≥97.0% (by GC, titration analysis)
Irritant
8-Mercaptomenthone ≥97.0% (by GC, titration analysis)
Catalog # TCIAM2023-5G
Supplier:  TCI
CAS Number:  
Irritant
8-Mercaptomenthone ≥97.0% (by GC, titration analysis)
Catalog # TCIAM2023-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    97.0 % (total of isomer)
  • Purity (Neutralization titration):
    min. 97.0 %

Specification Test Results

About this item

 :
 :186.31 g/mol
Boiling Pt:75°C
Flash Pt:108°C
EINECS:253-953-1
 :GHS07
CAS Number:38462-22-5
Formula:C₁₀H₁₈OS
MDL Number:MFCD00012393