
Bis(2-mercaptoethyl)sulphide ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3570-55-6
:
Formula:C₄H₁₀S₃
MDL Number:MFCD00004893
:154.30 g/mol
Boiling Pt:138°C
Flash Pt:90°C
EINECS:222-671-0
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %