
(2-Methyl-2-propenyl)succinic anhydride ≥97.0% (by GC, titration analysis)
Catalog # TCIAM1123-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Morpholine Method):min. 97.0 %
- Melting point:57.0 to 61.0 deg-C
- Solubility in Toluene:very faint turbidity
Specification Test Results
About this item
:
:154.17 g/mol
Boiling Pt:137°C
:GHS07
CAS Number:18908-20-8
Formula:C₈H₁₀O₃
MDL Number:MFCD00075477