

5-Methyl-2-heptene (cis- and trans- mixture) cis- and trans-mixture ≥95.0% (by GC)
: TCI
:22487-87-2
:
:C₈H₁₆
:MFCD00061016
:112.22 g/mol
:13°C
:3295
:3,II
:GHS02
:GHS08
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 % (total of isomers)
Frequently Bought Together