

Methyl propargyl ether ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:627-41-8
:
Formula:C₄H₆O
MDL Number:MFCD00008578
Boiling Pt:62°C
Flash Pt:–18°C
EINECS:210-998-1
UN:3271
ADR:3,II
:70.09 g/mol
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %