4-Methylesculetin ≥98.0% (by HPLC, titration analysis)
TCIAM0766-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Gray to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
:
:192.17 g/mol
:208-470-0
:GHS07
:529-84-0
:C₁₀H₈O₄
:MFCD00006859