N-Methyl-ε-caprolactam ≥97.0% (by GC)
: TCI
: N-Methyl-2-azacycloheptanone, 1-Methylazepan-2-one
:2556-73-2
:
:C₇H₁₃NO
:MFCD00003263
:127.19 g/mol
:14,05989
:120°C
:103°C
:219-874-1
:GHS07
- Environmentally Preferable:
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %