Order Entry
Finland
ContactUsLinkComponent
2-Methylbutyl-DL-2-methylbutyrate ≥97.0% (by GC)
2-Methylbutyl-DL-2-methylbutyrate ≥97.0% (by GC)
Catalog # TCIAM0578-25ML
Supplier:  TCI
CAS Number:  
2-Methylbutyl-DL-2-methylbutyrate ≥97.0% (by GC)
Catalog # TCIAM0578-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :172.27 g/mol
Boiling Pt:187°C
Flash Pt:67°C
EINECS:219-497-2
CAS Number:2445-78-5
Formula:C₁₀H₂₀O₂
MDL Number:MFCD00059395