

2-Methyl-1,5-heptadiene (cis- and trans- mixture) cis- and trans-mixture
Catalog # TCIAM0316-5ML
Supplier: TCI
CAS Number:
Specifications
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 % (total of isomers)
Specification Test Results
About this item
:
:110.20 g/mol
Boiling Pt:118°C
Flash Pt:14°C
UN:1993
ADR:3,II
:GHS02
:GHS08
CAS Number:6766-54-7
Formula:C₈H₁₄
MDL Number:MFCD00059240