Order Entry
Finland
ContactUsLinkComponent
5-Methyl-2-hexene (cis- and trans- mixture) cis- and trans-mixture ≥99.0% (by GC)
Flammable
Health
5-Methyl-2-hexene (cis- and trans- mixture) cis- and trans-mixture ≥99.0% (by GC)
Catalog # TCIAM0305-1ML
Supplier:  TCI
CAS Number:  
Flammable
Health
5-Methyl-2-hexene (cis- and trans- mixture) cis- and trans-mixture ≥99.0% (by GC)
Catalog # TCIAM0305-1ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Seq:
    2070

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 % (total of isomers)

Specification Test Results

About this item

 :
 :98.19 g/mol
UN:3295
ADR:3,II
 :GHS02
 :GHS08
CAS Number:3404-62-4
Formula:C₇H₁₄
MDL Number:MFCD00059239