Lithium phenyl(2,4,6-trimethylbenzoyl)phosphinate (LAP) ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:85073-19-4
:
Formula:C₁₆H₁₆LiO₃P
MDL Number:MFCD29472537
:294.21 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together