
Levofloxacin ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:100986-85-4
:
Formula:C₁₈H₂₀FN₃O₄
MDL Number:MFCD00865049
Merck Index:14,06771
:361.37 g/mol
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 96.0 %
- Melting point:222.0 to 230.0 deg-C
- Specific rotation [a]20/D:-63.0 to -70.0 deg (C=5, CHCl3)