SKF-96365 hydrochloride ≥98% (by HPLC)
TCIAK0066-50MG
: TCI
:
- Pk:50 mg
- Seq:2370
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):6.60 to 7.30 %
:
:402.9 g/mol
:119°C
:130495-35-1
:C₂₂H₂₆N₂O₃·HCl
:MFCD00236407