2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone ≥98.0% (by GC)
TCIAI1039-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Specific rotation [a]20/D:-65.0 to -70.0 deg (C=1, CHCL3)
:
:188.179 g/mol
:138°C
:30725-00-9
:C₈H₁₂O₅
:MFCD00080793