Order Entry
Finland
ContactUsLinkComponent
(Indenyl)titanium(IV) trichloride ≥98.0% (W)(T)
Corrosive
(Indenyl)titanium(IV) trichloride ≥98.0% (W)(T)
Catalog # TCIAI0646-1G
Supplier:  TCI
CAS Number:  
Corrosive
(Indenyl)titanium(IV) trichloride ≥98.0% (W)(T)
Catalog # TCIAI0646-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Dark green to Dark purple to Black powder to crystal
  • Purity (with ignition residue):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :269.37 g/mol
UN:3261
ADR:8,II
 :GHS05
CAS Number:84365-55-9
Formula:C₉H₇Cl₃Ti
MDL Number:MFCD00269849