



3-Isopropenyl-ɑ,ɑ-dimethylbenzyl isocyanate ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2094-99-7
:
Formula:C₁₃H₁₅NO
MDL Number:MFCD00080490
:201.27 g/mol
Boiling Pt:85°C
Flash Pt:124°C
Storage Temperature:Refrigerator
EINECS:402-440-2
UN:2206
ADR:6.1,II
:GHS05
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %