
Monoethyl itaconate ≥97.0% (by GC, titration analysis)
TCIAI0268-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:54.0 to 59.0 deg-C
- Solubility in Methanol:almost transparency
:
:158.15 g/mol
:153°C
:GHS07
:Refrigerator
:57718-07-7
:C₇H₁₀O₄
:MFCD00059138